Assay Details
| Gene Symbol: Mir6413 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_105840.1 | 1 | 55 | 90 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microtubule-associated protein 7 | Map7 |
E-MAP-115, MAP-7, Mtap7, R75000, mshi, mste, ste |
-16.0kb | Anti-sense | Chr. 10 - 20148920 - 20281590 on NCBI Build GRCm38 |
| Downstream | BCL2-associated transcription factor 1 | Bclaf1 |
2610102K23Rik, 2700025J07Rik, 2810454G14Rik, 5730534O06Rik, AI450190, AW556225, Btf, mKIAA0164 |
13.8kb | Anti-sense | Chr. 10 - 20311461 - 20342503 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir6413
Genomic Map