Assay Details
| Gene Symbol: Mir6418 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_105846.1 | 1 | 61 | 63 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | GRB10 interacting GYF protein 1 | Gigyf1 |
Perq1, mKIAA4110 |
-1.5kb | Anti-sense | Chr. 5 - 137510258 - 137527935 on NCBI Build GRCm38 |
| Downstream | actin-like 6B | Actl6b |
Actl6, ArpNa, Baf53b |
23.9kb | Anti-sense | Chr. 5 - 137553525 - 137569582 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir6418
Genomic Map