Assay Details
| Gene Symbol: Mir6987 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_105953.1 | 1 | 38 | 68 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | signal-induced proliferation associated gene 1 | Sipa1 | Spa1 | -15.1kb | Sense | Chr. 19 - 5651185 - 5663947 on NCBI Build GRCm38 |
| Downstream | mitogen-activated protein kinase kinase kinase 11 | Map3k11 |
2610017K16Rik, Mlk3, PTK1, SPRK |
9.7kb | Anti-sense | Chr. 19 - 5688740 - 5702865 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir6987
Genomic Map