Assay Details
| Gene Symbol: Mir7031 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_105998.1 | 1 | 29 | 57 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ataxin 2 | Atxn2 |
9630045M23Rik, ATX2, AW544490, Sca2 |
-4.3kb | Anti-sense | Chr. 5 - 121711609 - 121814950 on NCBI Build GRCm38 |
| Downstream |
family with sequence similarity 109, member A |
Fam109a |
A230106M15Rik, AU017694, IPIP27A, Ses1 |
29.7kb | Anti-sense | Chr. 5 - 121849000 - 121854606 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir7031
Genomic Map