Assay Details
| Gene Symbol: Mir7066 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_106033.1 | 1 | 30 | 61 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | neurocan | Ncan |
C230035B04, Cspg3, Cspg3-rs, Tgfbit |
Sense | Chr. 8 - 70093082 - 70120870 on NCBI Build GRCm38 | |
| Downstream | regulatory factor X-associated ankyrin-containing protein | Rfxank | Tvl1 | 28.2kb | Sense | Chr. 8 - 70130806 - 70139197 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir7066
Genomic Map