Assay Details
| Gene Symbol: Mir7075 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_106043.1 | 1 | 42 | 73 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | telomeric repeat binding factor 2 | Terf2 | TRF2 | Sense | Chr. 8 - 107069400 - 107096642 on NCBI Build GRCm38 | |
| Downstream | cytochrome b5 type B | Cyb5b |
1810044O22Rik, AU015618, Cyb5m |
54.6kb | Anti-sense | Chr. 8 - 107150661 - 107187470 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir7075
Genomic Map