Assay Details
| Gene Symbol: Mir7087 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_106055.1 | 1 | 35 | 55 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transgelin | Tagln |
Sm22, Sm22a, Ws310 |
-3.5kb | Sense | Chr. 9 - 45929628 - 45936058 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA A830035O19 gene | A830035O19Rik | ENSMUSG00000074379 | 15.1kb | Anti-sense | Chr. 9 - 45954730 - 45956000 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir7087
Genomic Map