Assay Details
| Gene Symbol: Mir7220 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_106079.1 | 1 | 33 | 57 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | arylsulfatase i | Arsi | BC020108 | -34.4kb | Sense | Chr. 18 - 60912240 - 60919504 on NCBI Build GRCm38 |
| Downstream | microRNA 6982 | Mir6982 |
Gm27890, mmu-mir-6982 |
4.1kb | Sense | Chr. 18 - 60958073 - 60958140 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir7220
Genomic Map