Assay Details
| Gene Symbol: Mir7222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_106081.1 | 1 | 39 | 57 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 7221 | Mir7221 |
Gm27378, mmu-mir-7221 |
-2.3kb | Sense | Chr. 2 - 92592206 - 92592277 on NCBI Build GRCm38 |
| Downstream | carbohydrate (keratan sulfate Gal-6) sulfotransferase 1 | Chst1 |
2610008E20Rik, AW125896, C6ST, GST-1, Gst1, KSGAL6ST |
5.0kb | Sense | Chr. 2 - 92599632 - 92615252 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir7222
Genomic Map