Assay Details
| Gene Symbol: Mir7242 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_106101.1 | 1 | 38 | 64 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 7241 | Mir7241 |
Gm27918, mmu-mir-7241 |
-1.7kb | Sense | Chr. 9 - 67727717 - 67727780 on NCBI Build GRCm38 |
| Downstream | uncharacterized LOC102633151 | LOC102633151 | 26.8kb | Sense | Chr. 9 - 67756420 - 67759516 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir7242
Genomic Map