Assay Details
| Gene Symbol: Mir7655 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_106115.1 | 1 | 29 | 53 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 10 |
Mllt10 |
Af10, B130021D15Rik, D630001B22Rik, mKIAA4140 |
Sense | Chr. 2 - 18055175 - 18212390 on NCBI Build GRCm38 | |
| Downstream |
DnaJ (Hsp40) homolog, subfamily C, member 1 |
Dnajc1 |
4733401K02Rik, AA960110, D230036H06Rik, Dnajl1, ERdj1, ERj1p, MTJ1 |
148.4kb | Anti-sense | Chr. 2 - 18206333 - 18392851 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir7655
Genomic Map