Assay Details
| Gene Symbol: Mir8107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_106186.1 | 1 | 59 | 71 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | neurobeachin-like 2 | Nbeal2 |
1110014F23Rik, BC042396, mKIAA0540 |
Sense | Chr. 9 - 110624789 - 110654161 on NCBI Build GRCm38 | |
| Downstream | coiled-coil domain containing 12 | Ccdc12 |
2700094L05Rik, C76605 |
20.4kb | Anti-sense | Chr. 9 - 110656503 - 110711593 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir8107
Genomic Map