Assay Details
| Gene Symbol: 1700065L07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_108032.1 | 4 - 5 | 392 | 93 | ||
| GenBank mRNA | AK006896.1 | - | 4 - 5 | 395 | 93 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
succinate-CoA ligase, GDP-forming, alpha subunit |
Suclg1 |
1500000I01Rik, Sucla1 |
-160.1kb | Anti-sense | Chr. 6 - 73248505 - 73276907 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 4931417E11 gene | 4931417E11Rik | 31.0kb | Sense | Chr. 6 - 73468573 - 73469667 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

1700065L07Rik
Genomic Map