Assay Details
| Gene Symbol: 4930463O16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_108059.1 | 1 - 2 | 888 | 105 | ||
| GenBank mRNA | AK015500.1 | - | 1 - 2 | 888 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

4930463O16Rik
Genomic Map