Assay Details
| Gene Symbol: 4930463O16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_108059.1 | 2 - 3 | 1291 | 86 | ||
| GenBank mRNA | AK015500.1 | - | 2 - 3 | 1291 | 86 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
NUAK family, SNF1-like kinase, 1 |
Nuak1 |
AU014801, AW494241, B230104P22Rik, Omphk1 |
-52.2kb | Anti-sense | Chr. 10 - 84370900 - 84440471 on NCBI Build GRCm38 |
| Downstream | cytoskeleton-associated protein 4 | Ckap4 |
5630400A09Rik, CLIMP-63, P63 |
32.4kb | Anti-sense | Chr. 10 - 84526305 - 84533888 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

4930463O16Rik
Genomic Map