Assay Details
| Gene Symbol: 9930111H07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_108086.1 | 3 - 4 | 193 | 116 | ||
| GenBank mRNA | AK020686.1 | - | 3 - 4 | 194 | 116 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 8096 | Mir8096 |
Gm27479, mmu-mir-8096 |
-29.4kb | Sense | Chr. 1 - 85751948 - 85752074 on NCBI Build GRCm38 |
| Downstream | calcium binding protein 39 | Cab39 |
AA408805, AA960512, C78372, MO25, MO25alpha |
8.7kb | Sense | Chr. 1 - 85793410 - 85851577 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

9930111H07Rik
Genomic Map