Assay Details
| Gene Symbol: Glb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_108101.1 | 1 - 2 | 193 | 99 | ||
| NR_108102.1 | 1 - 2 | 193 | 99 | |||
| GenBank mRNA | AK034966.1 | - | 164 | 99 | ||
| AK171760.1 | - | 1 - 2 | 164 | 99 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transmembrane protein with metallophosphoesterase domain | Tmppe | Sense | Chr. 9 - 114401095 - 114411201 on NCBI Build GRCm38 | ||
| Downstream | chemokine (C-C motif) receptor 4 | Ccr4 |
C-C CKR-4, CHEMR1, Cmkbr4, LESTR, Sdf1r |
69.8kb | Anti-sense | Chr. 9 - 114490316 - 114505473 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Glb1
Genomic Map