Assay Details
| Gene Symbol: 4631405J19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110333.1 | 3 - 4 | 500 | 62 | ||
| GenBank mRNA | BC100298.1 | - | 3 - 4 | 500 | 62 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | PR domain containing 11 | Prdm11 |
8030443D09Rik, AA163972, Gm707 |
Anti-sense | Chr. 2 - 92974906 - 93046144 on NCBI Build GRCm38 | |
| Downstream | transformation related protein 53 inducible protein 11 | Trp53i11 | Tp53i11 | 156.6kb | Sense | Chr. 2 - 93187534 - 93201757 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4631405J19Rik
Genomic Map