Assay Details
| Gene Symbol: Sh3yl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110343.1 | 2 - 3 | 707 | 84 | ||
| GenBank mRNA | AK168483.1 | - | 2 - 3 | 190 | 84 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
acid phosphatase 1, soluble |
Acp1 |
4632432E04Rik, AI427468, Acp-1, LMW-PTP |
-1.3kb | Anti-sense | Chr. 12 - 30893326 - 30911621 on NCBI Build GRCm38 |
| Downstream |
family with sequence similarity 110, member C |
Fam110c |
9030611O19Rik, AU019489 |
151.6kb | Sense | Chr. 12 - 31073968 - 31079940 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Sh3yl1
Genomic Map