Assay Details
| Gene Symbol: Adarb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110345.1 | 10 - 11 | 2657 | 64 | ||
| GenBank mRNA | BC059822.1 | - | 10 - 11 | 2610 | 6844585 | 64 |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
phosphofructokinase, platelet |
Pfkp |
1200015H23Rik, 9330125N24Rik, ATP-PFK, PFK-C, PFK-P |
-2.1mb | Anti-sense | Chr. 13 - 6579874 - 6648789 on NCBI Build GRCm38 |
| Downstream | WD repeat domain 37 | Wdr37 | 34.2kb | Anti-sense | Chr. 13 - 8802966 - 8871904 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Adarb2
Genomic Map