Assay Details
| Gene Symbol: Tbpl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110359.1 | 5 - 6 | 931 | 94 | ||
| GenBank mRNA | BC144905.1 | - | 5 - 6 | 909 | 94 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | histamine N-methyltransferase | Hnmt |
1500031F01Rik, AI788969 |
-26.2kb | Sense | Chr. 2 - 24002911 - 24049757 on NCBI Build GRCm38 |
| Downstream |
interleukin 1 family, member 8 |
Il1f8 |
2310043N20Rik, Il36b |
65.9kb | Anti-sense | Chr. 2 - 24153172 - 24160103 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Tbpl2
Genomic Map