Assay Details
| Gene Symbol: Eldr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110421.1 | 5 - 6 | 578 | 82 | ||
| NR_110423.1 | 3 - 4 | 442 | 82 | |||
| NR_110424.1 | 3 - 4 | 425 | 82 | |||
| GenBank mRNA | BC054406.1 | - | 3 - 4 | 186 | 82 | |
| BI990615.1 | - | 5 - 6 | 378 | 82 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | epidermal growth factor receptor | Egfr |
9030024J15Rik, AI552599, Erbb, Errb1, Errp, Wa5, wa-2, wa2 |
-20.8kb | Anti-sense | Chr. 11 - 16752203 - 16913907 on NCBI Build GRCm38 |
| Downstream | F-box protein 48 | Fbxo48 | A630050E13Rik | 14.8kb | Anti-sense | Chr. 11 - 16951375 - 16954777 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Eldr
Genomic Map