Assay Details
| Gene Symbol: A230103J11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110582.1 | 1 - 2 | 54 | 92 | ||
| GenBank mRNA | AK039159.1 | - | 1 - 2 | 55 | 92 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
transportin 2 (importin 3, karyopherin beta 2b) |
Tnpo2 |
1110034O24Rik, AA414969, AI464345, AI852433, IPO3, Knpb2b, Kpnb2b, TRN2 |
Anti-sense | Chr. 8 - 85036888 - 85057585 on NCBI Build GRCm38 | |
| Downstream | microRNA 7070 | Mir7070 |
Gm27565, mmu-mir-7070 |
1.6kb | Anti-sense | Chr. 8 - 85062086 - 85062171 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

A230103J11Rik
Genomic Map