Assay Details
| Gene Symbol: Ccdc169 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110776.1 | 7 - 8 | 600 | 71 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
spastic paraplegia 20, spartin (Troyer syndrome) homolog (human) |
Spg20 |
AI840044, C79168, SPARTIN, TAHCCP1, mKIAA0610 |
-26.0kb | Sense | Chr. 3 - 55112074 - 55137332 on NCBI Build GRCm38 |
| Downstream | spermatogenesis and oogenesis specific basic helix-loop-helix 2 | Sohlh2 |
4933406N12Rik, AI413378 |
11.6kb | Sense | Chr. 3 - 55182044 - 55209957 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Ccdc169
Genomic Map