Assay Details
| Gene Symbol: Ccdc169 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110780.1 | 6 - 7 | 416 | 76 | ||
| NR_110781.1 | 7 - 8 | 450 | 76 | |||
| GenBank mRNA | BC171976.1 | - | 6 - 7 | 369 | 76 | |
| BC171978.1 | - | 7 - 8 | 403 | 76 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
spastic paraplegia 20, spartin (Troyer syndrome) homolog (human) |
Spg20 |
AI840044, C79168, SPARTIN, TAHCCP1, mKIAA0610 |
-25.5kb | Sense | Chr. 3 - 55112074 - 55137332 on NCBI Build GRCm38 |
| Downstream | spermatogenesis and oogenesis specific basic helix-loop-helix 2 | Sohlh2 |
4933406N12Rik, AI413378 |
18.6kb | Sense | Chr. 3 - 55182044 - 55209957 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Ccdc169
Genomic Map