Assay Details
| Gene Symbol: Rp2h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110966.1 | 6 - 7 | 1319 | 108 | ||
| NR_110967.1 | 7 - 8 | 1456 | 108 | |||
| GenBank mRNA | AK044002.1 | - | 6 - 7 | 1226 | 108 | |
| AK076907.1 | - | 7 - 8 | 1318 | 108 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
solute carrier family 9 (sodium/hydrogen exchanger), member 7 |
Slc9a7 |
A530087D17Rik, NHE7 |
-112.2kb | Anti-sense | Chr. X - 20098851 - 20291970 on NCBI Build GRCm38 |
| Downstream | jade family PHD finger 3 | Jade3 |
5730598B06, AI851988, AW544806, Phf16, mKIAA0215 |
20.0kb | Sense | Chr. X - 20425327 - 20519939 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Rp2h
Genomic Map