Assay Details
| Gene Symbol: Ppp1r42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110971.1 | 6 - 7 | 837 | 100 | ||
| GenBank mRNA | AK015164.1 | - | 6 - 7 | 832 | 100 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transcription factor 24 | Tcf24 |
ENSMUSG00000073738, Gm10567, Gm2330 |
-186.0 | Sense | Chr. 1 - 9960163 - 9968436 on NCBI Build GRCm38 |
| Downstream |
COP9 (constitutive photomorphogenic) homolog, subunit 5 (Arabidopsis thaliana) |
Cops5 |
AI303502, CSN5, Jab1, Mov34, Sgn5 |
38.9kb | Sense | Chr. 1 - 10024600 - 10038159 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Ppp1r42
Genomic Map