Assay Details
| Gene Symbol: Tmem222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110978.1 | 1 - 2 | 256 | 74 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | synaptotagmin-like 1 | Sytl1 |
9030417L18, PSGL-1, Slp1 |
-6.7kb | Sense | Chr. 4 - 133253090 - 133263087 on NCBI Build GRCm38 |
| Downstream | WD and tetratricopeptide repeats 1 | Wdtc1 |
Gm695, adipose, adp |
14.7kb | Sense | Chr. 4 - 133292466 - 133353476 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Tmem222
Genomic Map