Assay Details
| Gene Symbol: Pla2g10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110990.1 | 4 - 5 | 987 | 80 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | bifunctional apoptosis regulator | Bfar |
3010001A07Rik, AI666707, AW107665, Bar, Rnf47 |
-11.4kb | Anti-sense | Chr. 16 - 13671858 - 13703612 on NCBI Build GRCm38 |
| Downstream |
phospholipase A2, group X, opposite strand |
Pla2g10os | 1810007I06Rik | 5.4kb | Anti-sense | Chr. 16 - 13729836 - 13739471 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Pla2g10
Genomic Map