Assay Details
| Gene Symbol: Dynlrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_111891.1 | 1 - 2 | 86 | 76 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
itchy, E3 ubiquitin protein ligase |
Itch |
6720481N21Rik, 8030492O04Rik, A130065M08, AIP4, C230047C07Rik |
-9.7kb | Sense | Chr. 2 - 155133379 - 155226855 on NCBI Build GRCm38 |
| Downstream | microtubule-associated protein 1 light chain 3 alpha | Map1lc3a |
1010001H21Rik, 4922501H04Rik, LC3, LC3a |
33.6kb | Sense | Chr. 2 - 155276364 - 155278073 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Dynlrb1
Genomic Map