Assay Details
| Gene Symbol: Lin9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120498.1 | 8 - 9 | 1036 | 98 | ||
| GenBank mRNA | BC088992.1 | - | 8 - 9 | 826 | 98 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
poly (ADP-ribose) polymerase family, member 1 |
Parp1 |
5830444G22Rik, AI893648, ARTD1, Adprp, Adprt1, C80510, PARP, PPOL, parp-1, sPARP-1 |
-66.1kb | Sense | Chr. 1 - 180568975 - 180601254 on NCBI Build GRCm38 |
| Downstream | Mix1 homeobox-like 1 (Xenopus laevis) | Mixl1 |
Mm1, Mml |
24.2kb | Anti-sense | Chr. 1 - 180693043 - 180709055 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Lin9
Genomic Map