Assay Details
| Gene Symbol: Slc6a17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_121629.1 | 11 - 12 | 3262 | 80 | ||
| GenBank mRNA | AK032262.1 | - | 11 - 12 | 3052 | 80 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
potassium voltage gated channel, Shaw-related subfamily, member 4 |
Kcnc4 |
AI850292, KSHIIIC, Kcr2-4, Kv3.4 |
-8.7kb | Sense | Chr. 3 - 107438296 - 107459952 on NCBI Build GRCm38 |
| Downstream | ubiquitin-like 4B | Ubl4b | 4930522D07Rik | 82.1kb | Sense | Chr. 3 - 107553698 - 107555073 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Slc6a17
Genomic Map