Assay Details
| Gene Symbol: Ppapdc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_121642.1 | 2 - 3 | 172 | 75 | ||
| NR_121643.1 | 2 - 3 | 172 | 75 | |||
| GenBank mRNA | BC023111.1 | - | 2 - 3 | 159 | 75 | |
| BC062173.1 | - | 2 - 3 | 145 | 75 | ||
| BC089556.1 | - | 2 - 3 | 156 | 75 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Wolf-Hirschhorn syndrome candidate 1-like 1 (human) | Whsc1l1 |
6720429E03, A530023P05, AI528490, NSD3, WHISTLE |
-600.0 | Sense | Chr. 8 - 25601175 - 25719667 on NCBI Build GRCm38 |
| Downstream | DDHD domain containing 2 | Ddhd2 |
2010305K11Rik, SAMWD1, mKIAA0725 |
4.7kb | Anti-sense | Chr. 8 - 25725324 - 25754590 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Ppapdc1b
Genomic Map