Assay Details
| Gene Symbol: Tmem161a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_122028.1 | 8 - 9 | 847 | 81 | ||
| GenBank mRNA | AK082758.1 | - | 8 - 9 | 843 | 81 | |
| AK167517.1 | - | 3 - 4 | 245 | 81 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | myocyte enhancer factor 2B | Mef2b | AI451606 | -13.2kb | Sense | Chr. 8 - 70152778 - 70167488 on NCBI Build GRCm38 |
| Downstream |
solute carrier family 25, member 42 |
Slc25a42 | 2900084M01Rik | 3.0kb | Anti-sense | Chr. 8 - 70184340 - 70212281 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Tmem161a
Genomic Map