Assay Details
| Gene Symbol: Sf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_122118.1 | 9 - 10 | 1665 | 61 | ||
| GenBank mRNA | AK152914.1 | - | 9 - 10 | 1444 | 61 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mitogen-activated protein kinase kinase kinase kinase 2 | Map4k2 |
AI385662, BL44, GCK, Rab8ip |
-18.4kb | Sense | Chr. 19 - 6341116 - 6355619 on NCBI Build GRCm38 |
| Downstream | muscle glycogen phosphorylase | Pygm |
AI115133, PG |
9.8kb | Sense | Chr. 19 - 6384429 - 6398459 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Sf1
Genomic Map