Assay Details
Gene Symbol: Hipk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_125731.1 | 9 - 10 | 2279 | 63 | ||
GenBank mRNA | AF071071.1 | - | 9 - 10 | 2279 | 63 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | olfactomedin-like 3 | Olfml3 |
2810002E22Rik, AI115209, AW550633, HNOEL-iso, ONT3, mONT3 |
-16.2kb | Sense | Chr. 3 - 103735394 - 103738001 on NCBI Build GRCm38 |
Downstream |
DNA cross-link repair 1B, PSO2 homolog (S. cerevisiae) |
Dclre1b |
AI452214, Apollo, SNMIB, mSNM1B |
43.9kb | Sense | Chr. 3 - 103800605 - 103809387 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
