Assay Details
| Gene Symbol: Sumo3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_125903.1 | 1 - 2 | 103 | 64 | ||
| GenBank mRNA | AA105565.1 | - | 1 - 2 | 103 | 64 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | pituitary tumor-transforming 1 interacting protein | Pttg1ip |
1810010L20Rik, AI314311, AU018448, C79540 |
-15.0kb | Sense | Chr. 10 - 77581767 - 77598732 on NCBI Build GRCm38 |
| Downstream | ubiquitin-conjugating enzyme E2G 2 | Ube2g2 |
1110003O05Rik, D10Xrf369, UBC7, Ubc7p |
8.3kb | Sense | Chr. 10 - 77622321 - 77645990 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Sumo3
Genomic Map