Assay Details
| Gene Symbol: E4f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK033285.1 | - | 5 - 6 | 759 | 109 | |
| AK077014.1 | - | 808 | 109 | |||
| AK144176.1 | - | 5 - 6 | 704 | 109 | ||
| BC005634.1 | - | 5 - 6 | 679 | 109 | ||
| BC046459.1 | - | 5 - 6 | 817 | 5322519 | 109 | |
| BC071228.1 | - | 5 - 6 | 760 | 4915902 | 109 | |
| X76858.1 | - | 4 - 5 | 543 | 109 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | deoxyribonuclease 1-like 2 | Dnase1l2 | 4733401H14Rik | -3.0kb | Sense | Chr. 17 - 24440087 - 24443765 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 4833447I15 gene | 4833447I15Rik | 14.0kb | Sense | Chr. 17 - 24461220 - 24470191 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

E4f1
Genomic Map