Assay Details
| Gene Symbol: 4930481A15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027938.2 | 1 - 2 | 265 | 65 | ||
| GenBank mRNA | BI202508.1 | - | 2 - 3 | 65 | ||
| BM937611.1 | - | 1 - 2 | 194 | 65 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | testis specific 10 interacting protein | Tsga10ip |
1810013N20Rik, Fam161c |
-4.3kb | Anti-sense | Chr. 19 - 5390047 - 5402534 on NCBI Build GRCm38 |
| Downstream | Dr1 associated protein 1 (negative cofactor 2 alpha) | Drap1 |
2310074H19Rik, NC2-alpha |
7.0kb | Anti-sense | Chr. 19 - 5422804 - 5424979 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC0100003051.rn
Set Membership: |
|

4930481A15Rik
Genomic Map