Assay Details
| Gene Symbol: Gm7977 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040408.1 | 1 | 518 | 101 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 6379 | Mir6379 |
Gm27730, mmu-mir-6379 |
-1.1mb | Sense | Chr. 3 - 46900668 - 46900782 on NCBI Build GRCm38 |
| Downstream | enhancer of rudimentary homolog pseudogene | 1700018B24Rik | 579.0kb | Sense | Chr. 3 - 48605731 - 48609100 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm7977
Genomic Map