Assay Details
| Gene Symbol: Mir1291 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_106171.1 | 1 | 63 | 74 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
small nucleolar RNA, H/ACA box 34 |
Snora34 | MBI-89 | Sense | Chr. 15 - 98519717 - 98519834 on NCBI Build GRCm38 | |
| Downstream |
small nucleolar RNA, H/ACA box 2B |
Snora2b | MBI-137 | 6.5kb | Sense | Chr. 15 - 98526348 - 98526459 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay targets the same location as MTA transcript cluster(s) TC1500002177.mm
This assay targets the same location as RTA transcript cluster(s) TC1000002356.rn
This assay targets the same location as RTA transcript cluster(s) TC1000002356.rn
Set Membership: |
|

Mir1291
Genomic Map