Assay Details
| Gene Symbol: Gm24148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_106184.1 | 1 | 92 | 104 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
small nucleolar RNA, H/ACA box 75 |
Snora75 | MBI-55 | -696.0 | Sense | Chr. 1 - 86351170 - 86351285 on NCBI Build GRCm38 |
| Downstream |
small nucleolar RNA, C/D box 82 |
Snord82 |
Rnu82, U82, Z25 |
4.1kb | Sense | Chr. 1 - 86356260 - 86356327 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gm24148
Genomic Map