Assay Details
| Gene Symbol: Mir6390 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_105814.1 | 1 | 76 | 115 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 9330188P03 gene | 9330188P03Rik |
Gm1589, mgt-6 |
-29.7kb | Anti-sense | Chr. 14 - 105589549 - 105593929 on NCBI Build GRCm38 |
| Downstream | sprouty homolog 2 (Drosophila) | Spry2 | sprouty2 | 268.2kb | Sense | Chr. 14 - 105891947 - 105896819 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir6390
Genomic Map