Assay Details
| Gene Symbol: Krt86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010667.2 | NP_034797.1 | 8 - 9 | 1350 | 68 | |
| GenBank mRNA | AK160496.1 | - | 2 - 3 | 219 | 68 | |
| AK161149.1 | - | 8 - 9 | 1352 | 68 | ||
| BC139222.1 | - | 8 - 9 | 1328 | 8862244 | 68 | |
| X99143.1 | - | 8 - 9 | 1187 | 68 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Krt86
Genomic Map