Assay Details
| Gene Symbol: Nid2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008695.2 | NP_032721.2 | 17 - 18 | 3886 | 71 | |
| GenBank mRNA | AB017202.1 | - | 17 - 18 | 3848 | 71 | |
| AK045306.1 | - | 17 - 18 | 3861 | 71 | ||
| AK051664.1 | - | 17 - 18 | 3866 | 71 | ||
| AK083629.1 | - | 17 - 18 | 3863 | 71 | ||
| AK140832.1 | - | 17 - 18 | 3860 | 71 | ||
| AK164258.1 | - | 17 - 18 | 3857 | 71 | ||
| AY074820.1 | - | 17 - 18 | 3847 | 71 | ||
| BC054746.1 | - | 17 - 18 | 3868 | 6405073 | 71 | |
| BC057016.1 | - | 17 - 18 | 3814 | 5719452 | 71 | |
More Information
Set Membership: |
|

Nid2
Genomic Map