Assay Details
| Gene Symbol: Nars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153591.4 | NP_705819.3 | 6 - 7 | 964 | 75 | |
| GenBank mRNA | AK046799.1 | - | 6 - 7 | 965 | 75 | |
| AK046801.1 | - | 6 - 7 | 965 | 75 | ||
| AK047091.1 | - | 6 - 7 | 909 | 75 | ||
| AK050718.1 | - | 6 - 7 | 903 | 75 | ||
| AK138260.1 | - | 6 - 7 | 944 | 75 | ||
| AK164179.1 | - | 6 - 7 | 933 | 75 | ||
| AK189381.1 | - | 2 - 3 | 115 | 75 | ||
| BC030041.1 | - | 2 - 3 | 168 | 1226297 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC030041.1 transcripts.
Set Membership: |
|

Nars2
Genomic Map