Assay Details
| Gene Symbol: Scn3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018732.3 | NP_061202.3 | 1 - 2 | 226 | 91 | |
| GenBank mRNA | AK148881.1 | - | 1 - 2 | 225 | 91 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK148881.1, NM_018732.3 transcripts.
Set Membership: |
|

Scn3a
Genomic Map