Assay Details
| Gene Symbol: 1700065D16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271569.1 | NP_001258498.1 | 1 - 2 | 378 | 67 | |
| GenBank mRNA | BU945830.1 | - | 313 | 67 | ||
| CN834250.1 | - | 1 - 2 | 370 | 67 | ||
More Information
Set Membership: |
|

1700065D16Rik
Genomic Map