Assay Details
| Gene Symbol: Slc7a15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286385.1 | NP_001273314.1 | 1 - 2 | 777 | 75 | |
| GenBank mRNA | AK008024.1 | - | 1 - 2 | 778 | 75 | |
More Information
Set Membership: |
|

Slc7a15
Genomic Map