Assay Details
| Gene Symbol: Mapk7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291033.1 | NP_001277962.1 | 5 - 6 | 2442 | 93 | |
| NM_001291034.1 | NP_001277963.1 | 4 - 5 | 2517 | 93 | ||
| NM_001291035.1 | NP_001277964.1 | 4 - 5 | 2658 | 93 | ||
| NM_001291036.1 | NP_001277965.1 | 5 - 6 | 2507 | 93 | ||
| NM_001291037.1 | NP_001277966.1 | 4 - 5 | 2657 | 93 | ||
| NM_011841.2 | NP_035971.1 | 5 - 6 | 2392 | 93 | ||
| GenBank mRNA | AB019373.1 | - | 5 - 6 | 2342 | 93 | |
| AF126159.1 | - | 4 - 5 | 2155 | 93 | ||
| AF126160.1 | - | 4 - 5 | 2205 | 93 | ||
| AF126161.1 | - | 4 - 5 | 2575 | 93 | ||
| AK148119.1 | - | 5 - 6 | 2386 | 93 | ||
| AK155187.1 | - | 5 - 6 | 2322 | 93 | ||
| AY534740.1 | - | 4 - 5 | 2429 | 93 | ||
| BC013745.1 | - | 4 - 5 | 2588 | 93 | ||
| BC033598.1 | - | 5 - 6 | 2482 | 93 | ||
| BC100398.1 | - | 4 - 5 | 2459 | 30470655 | 93 | |
More Information
Set Membership: |
|

Mapk7
Genomic Map